missing translation for 'onlineSavingsMsg'
Learn More

2-Isobutyl-3-methoxypyrazine, 98%

Catalog No. AAA1368006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1368006 5 g
AAA1368003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1368006 Supplier Thermo Scientific Chemicals Supplier No. A1368006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 24683-00-9 | C9H14N2O | 166.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 24683-00-9
Molecular Formula C9H14N2O
Molecular Weight (g/mol) 166.22
MDL Number MFCD00006128
InChI Key UXFSPRAGHGMRSQ-UHFFFAOYSA-N
Synonym 2-isobutyl-3-methoxypyrazine, 3-isobutyl-2-methoxypyrazine, 2-methoxy-3-2-methylpropyl pyrazine, 2-methoxy-3-isobutylpyrazine, 2-methoxy-3-isobutyl pyrazine, 1dzk, pyrazine, 2-methoxy-3-2-methylpropyl, unii-s327o0t12o, ibmp, 3-methoxy-2-isobutylpyrazine
PubChem CID 32594
IUPAC Name 2-methoxy-3-(2-methylpropyl)pyrazine
SMILES COC1=NC=CN=C1CC(C)C

Specifications

Density 0.986
Flash Point 80°C (176°F)
Refractive Index 1.49
Quantity 5 g
Formula Weight 166.22
Percent Purity 98%
Chemical Name or Material 2-Isobutyl-3-methoxypyrazine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 246-402-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only