Learn More
2-Iodoxybenzoic acid, stabilized
CAS: 61717-82-6 | C7H5IO4 | 280.01 g/mol
$365.22 - $1231.32
Chemical Identifiers
| CAS | 61717-82-6 |
|---|---|
| Molecular Formula | C7H5IO4 |
| Molecular Weight (g/mol) | 280.01 |
| MDL Number | MFCD00024166 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC374650100
|
Thermo Scientific Chemicals
374650100 |
10 g | Glass bottle |
Each for $365.22
|
|
||||
|
AC374650500
|
Thermo Scientific Chemicals
374650500 |
50 g | Glass bottle |
Each for $1,231.32
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
oxidation reagentChemical Identifiers
| 61717-82-6 | |
| C7H5IO4 | |
| 280.01 | |
| MFCD00024166 |
Specifications
| 61717-82-6,64297-64-9,121-91-5,65-85-0 | |
| White | |
| C7H5IO4 | |
| 10 g | |
| Solubility in water: slightly soluble | |
| 280.01 | |
| Powder, Prills or Agglomerates |
| 250.0°C | |
| Glass bottle | |
| MFCD00024166 | |
| 15, 5091 | |
| 280.01 | |
| 39 to 50% as 2-Iodoxybenzoic acid | |
| 2-Iodoxybenzoic acid |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Causes damage to organs through prolonged or repeated exposure if inhaled.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
RUO – Research Use Only