missing translation for 'onlineSavingsMsg'
Learn More

2-Iodoxybenzoic Acid (stabilized with Benzoic Acid + Isophthalic Acid) 39.0+%, TCI America™

Catalog No. p-7131598
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I07915G 5 g
I079125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I07915G Supplier TCI America Supplier No. I07915G
Only null left

Chemical Identifiers

CAS 61717-82-6
Molecular Formula C7H5IO4
MDL Number MFCD02912492
Synonym IBX

Specifications

Melting Point 232°C
Color White
Quantity 5 g
UN Number 1759
Formula Weight 280.02
Percent Purity ≥39.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Iodoxybenzoic Acid (stabilized with Benzoic Acid + Isophthalic Acid)