missing translation for 'onlineSavingsMsg'
Learn More

2-Iodosobenzoic acid, 98%, Thermo Scientific™

Catalog No. AC446030050
Change view
Click to view available options
Quantity:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC446030050 5g
AC446030010 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC446030050 Supplier Thermo Scientific Chemicals Supplier No. 446030050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

oxidation reagent

Chemical Identifiers

CAS 304-91-6
Molecular Formula C7H5IO3
Molecular Weight (g/mol) 264.02
InChI Key IFPHDUVGLXEIOQ-UHFFFAOYSA-N
Synonym 2-iodosobenzoic acid, o-iodosobenzoic acid, o-iodosylbenzoic acid, o-iodosobenzoate, benzoic acid, 2-iodosyl, benzoic acid, o-iodoso, 2-iodosyl benzoate, benzoic acid, iodosyl, ortho-iodosylbenzoic acid, 3h-1,2-benziodoxol-1-ium, 3-oxo-, hydroxide
PubChem CID 67537
ChEBI CHEBI:52698
IUPAC Name 2-iodosylbenzoic acid
SMILES C1=CC=C(C(=C1)C(=O)O)I=O

Specifications

Infrared Spectrum Authentic
Assay Percent Range 97.5 to 102.5% (Iodometry)
Quantity 5g
Formula Weight 264.02
Percent Purity 98%
Chemical Name or Material 2-Iodosobenzoic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 206-159-4

RUO – Research Use Only