missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Iodopyridine (stabilized with Na2S2O3) 98.0+%, TCI America™
$111.66 - $367.70
Chemical Identifiers
| CAS | 5029-67-4 |
|---|---|
| Molecular Formula | C5H4IN |
| Molecular Weight (g/mol) | 205.00 |
| MDL Number | MFCD00464928 |
| InChI Key | CCZWSTFVHJPCEM-UHFFFAOYSA-N |
| Synonym | 2-iodo-pyridine, pyridine, iodo, iodopyridine, 2-iodo pyridine, 2-iodopyridine, 2-pyridyl iodide, 2-pyridyl iodide;, pyridine, 2-iodo, pubchem4074, acmc-1avou |
| PubChem CID | 221126 |
| IUPAC Name | 2-iodopyridine |
| SMILES | IC1=CC=CC=N1 |
Chemical Identifiers
| 5029-67-4 | |
| 205.00 | |
| CCZWSTFVHJPCEM-UHFFFAOYSA-N | |
| 221126 | |
| IC1=CC=CC=N1 |
| C5H4IN | |
| MFCD00464928 | |
| 2-iodo-pyridine, pyridine, iodo, iodopyridine, 2-iodo pyridine, 2-iodopyridine, 2-pyridyl iodide, 2-pyridyl iodide;, pyridine, 2-iodo, pubchem4074, acmc-1avou | |
| 2-iodopyridine |
Specifications
| 5029-67-4 | |
| 102°C | |
| MFCD00464928 | |
| 2-iodo-pyridine, pyridine, iodo, iodopyridine, 2-iodo pyridine, 2-iodopyridine, 2-pyridyl iodide, 2-pyridyl iodide;, pyridine, 2-iodo, pubchem4074, acmc-1avou | |
| IC1=CC=CC=N1 | |
| 205.00 | |
| 205.00 | |
| Liquid |
| Yellow | |
| C5H4IN | |
| 5 g | |
| CCZWSTFVHJPCEM-UHFFFAOYSA-N | |
| 2-iodopyridine | |
| 221126 | |
| ≥98.0% (GC) | |
| 2-Iodopyridine (stabilized with Na2S2O3) |
Safety and Handling
TSCA : No