missing translation for 'onlineSavingsMsg'
Learn More

2-Iodopyridine, 95%

CAS: 5029-67-4 | C5H4IN | 205.00 g/mol

$141.05 - $579.01

Chemical Identifiers

CAS 5029-67-4
Molecular Formula C5H4IN
Molecular Weight (g/mol) 205.00
MDL Number MFCD00464928
InChI Key CCZWSTFVHJPCEM-UHFFFAOYSA-N
Synonym 2-iodo-pyridine, pyridine, iodo, iodopyridine, 2-iodo pyridine, 2-iodopyridine, 2-pyridyl iodide, 2-pyridyl iodide;, pyridine, 2-iodo, pubchem4074, acmc-1avou
PubChem CID 221126
IUPAC Name 2-iodopyridine
SMILES IC1=CC=CC=N1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6306206
View Documents
Thermo Scientific Chemicals
H6306206
5 g
Each for $141.05
Only null left
 
AAH6306214
View Documents
Thermo Scientific Chemicals
H6306214
25 g
Each for $579.01
Only null left
 
Description

Description

2-Iodopyridine is a reagent used in the preparation of human NAD+-dependent 15-hydroxyprostaglandin dehydrogenase inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Iodopyridine is a reagent used in the preparation of human NAD+-dependent 15-hydroxyprostaglandin dehydrogenase inhibitors.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Specifications

Chemical Identifiers

5029-67-4
205.00
CCZWSTFVHJPCEM-UHFFFAOYSA-N
221126
IC1=CC=CC=N1
C5H4IN
MFCD00464928
2-iodo-pyridine, pyridine, iodo, iodopyridine, 2-iodo pyridine, 2-iodopyridine, 2-pyridyl iodide, 2-pyridyl iodide;, pyridine, 2-iodo, pubchem4074, acmc-1avou
2-iodopyridine

Specifications

5029-67-4
Yellow
C5H4IN
MFCD00464928
105790
2-iodo-pyridine, pyridine, iodo, iodopyridine, 2-iodo pyridine, 2-iodopyridine, 2-pyridyl iodide, 2-pyridyl iodide;, pyridine, 2-iodo, pubchem4074, acmc-1avou
CCZWSTFVHJPCEM-UHFFFAOYSA-N
2-iodopyridine
221126
95%
2-Iodopyridine
52°C (decomposition)
1.928 g/mL
1.632
5 g
Light sensitive
Slightly soluble in water.
IC1=CC=CC=N1
205.00
205
Solid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only