missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Iodopyridine, 95%
Description
2-Iodopyridine is a reagent used in the preparation of human NAD+-dependent 15-hydroxyprostaglandin dehydrogenase inhibitors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Iodopyridine is a reagent used in the preparation of human NAD+-dependent 15-hydroxyprostaglandin dehydrogenase inhibitors.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
2-Iodopyridine is a reagent used in the preparation of human NAD+-dependent 15-hydroxyprostaglandin dehydrogenase inhibitors.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Specifications
Specifications
| Melting Point | 52°C (decomposition) |
| Color | Yellow |
| Density | 1.928 g/mL |
| Refractive Index | 1.632 |
| Quantity | 25 g |
| Beilstein | 105790 |
| Sensitivity | Light sensitive |
| Solubility Information | Slightly soluble in water. |
| Formula Weight | 205 |
| Percent Purity | 95% |
| Show More |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only