missing translation for 'onlineSavingsMsg'
Learn More

2-Iodopyridine, 95%

Catalog No. AAH6306206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6306206 5 g
AAH6306214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6306206 Supplier Thermo Scientific Chemicals Supplier No. H6306206

CAS: 5029-67-4 | C5H4IN | 205.00 g/mol

2-Iodopyridine is a reagent used in the preparation of human NAD+-dependent 15-hydroxyprostaglandin dehydrogenase inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Iodopyridine is a reagent used in the preparation of human NAD+-dependent 15-hydroxyprostaglandin dehydrogenase inhibitors.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.

Chemical Identifiers

CAS 5029-67-4
Molecular Formula C5H4IN
Molecular Weight (g/mol) 205.00
MDL Number MFCD00464928
InChI Key CCZWSTFVHJPCEM-UHFFFAOYSA-N
Synonym 2-iodo-pyridine, pyridine, iodo, iodopyridine, 2-iodo pyridine, 2-iodopyridine, 2-pyridyl iodide, 2-pyridyl iodide;, pyridine, 2-iodo, pubchem4074, acmc-1avou
PubChem CID 221126
IUPAC Name 2-iodopyridine
SMILES IC1=CC=CC=N1

Specifications

Melting Point 52°C (decomposition)
Color Yellow
Density 1.928 g/mL
Refractive Index 1.632
Quantity 5 g
Beilstein 105790
Sensitivity Light sensitive
Solubility Information Slightly soluble in water.
Formula Weight 205
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-Iodopyridine
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only