missing translation for 'onlineSavingsMsg'
Learn More

2-Iodopyrazine, 95%

Catalog No. AAH2624403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2624403 1 g
AAH2624406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2624403 Supplier Thermo Scientific Chemicals Supplier No. H2624403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 32111-21-0 | C4H3IN2 | 205.986 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 32111-21-0
Molecular Formula C4H3IN2
Molecular Weight (g/mol) 205.986
MDL Number MFCD01319019
InChI Key OYWPFIUVDKHHGQ-UHFFFAOYSA-N
Synonym iodopyrazine, 2-iodo-pyrazine, 2-iodo-1,3-diazine, iodo-pyrazine, 2-iodo pyrazine, pyrazine, 2-iodo, acmc-1ad2s
PubChem CID 642841
IUPAC Name 2-iodopyrazine
SMILES C1=CN=C(C=N1)I

Specifications

Density 2.086
Boiling Point 109°C to 110°C (34 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.655
Quantity 1 g
Sensitivity Light sensitive
Formula Weight 205.98
Percent Purity 95%
Chemical Name or Material 2-Iodopyrazine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only