missing translation for 'onlineSavingsMsg'
Learn More

2-Iodopropane (stabilized with Copper chip) 99.0+%, TCI America™

Catalog No. I006925G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I006925G 25 g
I0069100G 100 g
I0069500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I006925G Supplier TCI America Supplier No. I006925G

Chemical Identifiers

CAS 75-30-9
Molecular Formula C3H7I
Molecular Weight (g/mol) 169.993
MDL Number MFCD00001071
InChI Key FMKOJHQHASLBPH-UHFFFAOYSA-N
Synonym isopropyl iodide, propane, 2-iodo, 2-propyl iodide, sec-propyl iodide, isopropyliodide, 2-jodpropan, 2-jodpropan czech, 2-iodo-propane, i-propyl iodide, unii-67k05opz0e
PubChem CID 6362
IUPAC Name 2-iodopropane
SMILES CC(C)I

Specifications

Melting Point -90°C
Color Yellow
Boiling Point 90°C
Quantity 25 g
UN Number 2392
Formula Weight 169.99
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Iodopropane (stabilized with Copper chip)
EINECSNumber (2)-3788
RTECSNumber TZ4200000
TSCA Yes