missing translation for 'onlineSavingsMsg'
Learn More

2-Iodophenylacetic acid, 97+%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 394040050

Quantity 5g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC394040050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

18698-96-9
262.04
IUHXGZHKSYYDIL-UHFFFAOYSA-N
2780090
C1=CC=C(C(=C1)CC(=O)O)I
C8H7IO2
MFCD00046546
2-iodophenylacetic acid, 2-2-iodophenyl acetic acid, benzeneacetic acid, 2-iodo, 2-iodophenyl acetic acid, o-iodophenylacetic acid, 2-iodobenzeneacetic acid, 2-iodophenylaceticacid, pubchem2552, acmc-209eox, maybridge1_005605
2-(2-iodophenyl)acetic acid

Specifications

2-Iodophenylacetic acid
97.0
Authentic
Glass bottle
IC6H4CH2CO2H
MFCD00046546
IUHXGZHKSYYDIL-UHFFFAOYSA-N
2-(2-iodophenyl)acetic acid
2780090
97+%
18698-96-9
100.0
97% min. (HPLC)
C8H7IO2
5g
2-iodophenylacetic acid, 2-2-iodophenyl acetic acid, benzeneacetic acid, 2-iodo, 2-iodophenyl acetic acid, o-iodophenylacetic acid, 2-iodobenzeneacetic acid, 2-iodophenylaceticacid, pubchem2552, acmc-209eox, maybridge1_005605
C1=CC=C(C(=C1)CC(=O)O)I
262.04
262.04
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

Promotions

Promotions

Promo
Expires: 06-30-2027
Get Up to $1,800 in Chemicals with Purchase
Spend $250 or more on Thermo Fisher Scientific chemicals and get up to $1,800 to spend on additional chemicals.

Adding...