missing translation for 'onlineSavingsMsg'
Learn More

2-Iodophenylacetic acid, 97+%, Thermo Scientific™

Catalog No. AC394040010
Change view
Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC394040010 1g Glass bottle
AC394040050 5g Glass bottle
AC394040250 25g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC394040010 Supplier Thermo Scientific Chemicals Supplier No. 394040010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 18698-96-9
Molecular Formula C8H7IO2
Molecular Weight (g/mol) 262.04
MDL Number MFCD00046546
InChI Key IUHXGZHKSYYDIL-UHFFFAOYSA-N
Synonym 2-iodophenylacetic acid, 2-2-iodophenyl acetic acid, benzeneacetic acid, 2-iodo, 2-iodophenyl acetic acid, o-iodophenylacetic acid, 2-iodobenzeneacetic acid, 2-iodophenylaceticacid, pubchem2552, acmc-209eox, maybridge1_005605
PubChem CID 2780090
IUPAC Name 2-(2-iodophenyl)acetic acid
SMILES C1=CC=C(C(=C1)CC(=O)O)I

Specifications

CAS Min % 97.0
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 97% min. (HPLC)
Packaging Glass bottle
Linear Formula IC6H4CH2CO2H
Quantity 1g
Formula Weight 262.04
Percent Purity 97+%
Chemical Name or Material 2-Iodophenylacetic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only