missing translation for 'onlineSavingsMsg'
Learn More

2-Iodoimidazole, 96%, Thermo Scientific Chemicals

Catalog No. AAH5186803
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5186803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5186803 Supplier Thermo Scientific Chemicals Supplier No. H5186803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3034-62-6 | Molecular Formula: C3H3IN2 | Molecular Weight: 193.975 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3034-62-6
Molecular Formula C3H3IN2
Molecular Weight (g/mol) 193.975
MDL Number MFCD00159701
InChI Key MHHDMDLNVVCTAJ-UHFFFAOYSA-N
Synonym 2-iodoimidazole, iodoimidazole, 1h-imidazole, 2-iodo, sftheavililuh@, 2-iodoimidazole hcl, pubchem9423, acmc-1aies, 2-iodo imidazole
PubChem CID 2773468
IUPAC Name 2-iodo-1H-imidazole
SMILES C1=CN=C(N1)I

Specifications

Melting Point 190°C to 191°C
Quantity 1 g
Sensitivity Light sensitive
Formula Weight 193.96
Percent Purity 96%
Chemical Name or Material 2-Iodoimidazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only