missing translation for 'onlineSavingsMsg'
Learn More

(2-Iodoethyl)benzene (stabilized with Copper chip) 98.0+%, TCI America™

Catalog No. p-7131457
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I05215G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I05215G Supplier TCI America Supplier No. I05215G
Only null left

Chemical Identifiers

CAS 17376-04-4
Molecular Formula C8H9I
Molecular Weight (g/mol) 232.064
MDL Number MFCD00039408
InChI Key KVTHPKXDLVYNCH-UHFFFAOYSA-N
Synonym 2-iodoethyl benzene, benzene, 2-iodoethyl, 2-phenylethyl iodide, phenethyl iodide, iodoethyl benzene, .beta.-phenethyl iodide, phenethyliodide, p-iodoethylbenzene, phenylethyl iodide, 4-iodoethylbenzene
PubChem CID 28503
IUPAC Name 2-iodoethylbenzene
SMILES C1=CC=C(C=C1)CCI

Specifications

Boiling Point 123°C
Quantity 5 g
Formula Weight 232.06
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (2-Iodoethyl)benzene (stabilized with Copper chip)