missing translation for 'onlineSavingsMsg'
Learn More

2-Iodoethanol (stabilized with Copper chip) 98.0+%, TCI America™

Catalog No. I03915G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I03915G 5 g
I039125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I03915G Supplier TCI America Supplier No. I03915G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 624-76-0
Molecular Formula C2H5IO
Molecular Weight (g/mol) 171.97
MDL Number MFCD00002830
InChI Key QSECPQCFCWVBKM-UHFFFAOYSA-N
Synonym ethanol, 2-iodo, iodoethanol, ethylene iodohydrin, 2-iodoethan-1-ol, iodoethyl alcohol, 2-indoethanol, 2-iodo ethanol, 2-iodo-ethanol, 2-iodo-1-ethanol, 2-iodoethyl alcohol
PubChem CID 12225
IUPAC Name 2-iodoethan-1-ol
SMILES OCCI

Specifications

Color Yellow
Boiling Point 85°C
Quantity 5 g
UN Number 2810
Formula Weight 171.97
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Iodoethanol (stabilized with Copper chip)
RTECSNumber KL3820000
TSCA Yes
Recommended Storage Refrigerator