missing translation for 'onlineSavingsMsg'
Learn More

2-Iodoethanol, 99%, stabilized

Catalog No. p-4510613
Change view
Click to view available options
Quantity:
10 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC175500100 10 mL
AC175500500 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC175500100 Supplier Thermo Scientific Chemicals Supplier No. 175500100

CAS: 624-76-0 | C2H5IO | 171.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 624-76-0
Molecular Formula C2H5IO
Molecular Weight (g/mol) 171.97
MDL Number MFCD00002830
InChI Key QSECPQCFCWVBKM-UHFFFAOYSA-N
Synonym ethanol, 2-iodo, iodoethanol, ethylene iodohydrin, 2-iodoethan-1-ol, iodoethyl alcohol, 2-indoethanol, 2-iodo ethanol, 2-iodo-ethanol, 2-iodo-1-ethanol, 2-iodoethyl alcohol
PubChem CID 12225
SMILES OCCI

Specifications

Density 2.2050g/mL
Boiling Point 172.0°C to 175.0°C
Flash Point 65°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5690 to 1.5720
Linear Formula ICH2CH2OH
Quantity 10 mL
Beilstein 01,339
Fieser 13,348
Specific Gravity 2.205
IUPAC Name 2-iodoethanol
Stabilizer Copper
Formula Weight 171.97
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Iodoethanol
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Fatal if swallowed.
Toxic if swallowed.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wash face,hands and any exposed skin thoroughly after handling.
Rinse mouth.
EINECSNumber 210-861-6

RUO – Research Use Only