missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Iododibenzofuran 98.0+%, TCI America™
$410.07 - $1218.64
Chemical Identifiers
| CAS | 5408-56-0 |
|---|---|
| Molecular Formula | C12H7IO |
| Molecular Weight (g/mol) | 294.091 |
| MDL Number | MFCD00092649 |
| InChI Key | MNKSJEJNCQWHFE-UHFFFAOYSA-N |
| PubChem CID | 223327 |
| IUPAC Name | 2-iododibenzofuran |
| SMILES | C1=CC=C2C(=C1)C3=C(O2)C=CC(=C3)I |
Chemical Identifiers
| 5408-56-0 | |
| 294.091 | |
| MNKSJEJNCQWHFE-UHFFFAOYSA-N | |
| 2-iododibenzofuran |
| C12H7IO | |
| MFCD00092649 | |
| 223327 | |
| C1=CC=C2C(=C1)C3=C(O2)C=CC(=C3)I |
Specifications
| 5408-56-0 | |
| C12H7IO | |
| 1 g | |
| C1=CC=C2C(=C1)C3=C(O2)C=CC(=C3)I | |
| 294.091 | |
| 294.09 | |
| Crystalline Powder |
| 112°C | |
| MFCD00092649 | |
| MNKSJEJNCQWHFE-UHFFFAOYSA-N | |
| 2-iododibenzofuran | |
| 223327 | |
| ≥98.0% (GC) | |
| 2-Iododibenzofuran |
Safety and Handling
TSCA : No