missing translation for 'onlineSavingsMsg'
Learn More

2-Iodobenzenesulfonyl Chloride 98.0+%, TCI America™

$165.90 - $497.70

Chemical Identifiers

CAS 63059-29-0
Molecular Formula C6H4ClIO2S
Molecular Weight (g/mol) 302.51
MDL Number MFCD00833336
InChI Key ZPNWNLNPDRYULE-UHFFFAOYSA-N
PubChem CID 627400
IUPAC Name 2-iodobenzene-1-sulfonyl chloride
SMILES ClS(=O)(=O)C1=CC=CC=C1I
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I08111G
View Documents
TCI America
I08111G
1 g
Each for $165.90
Only null left
 
I08115G
View Documents
TCI America
I08115G
5 g
Each for $497.70
Only null left
 
Description

Description

Specifications

Chemical Identifiers

63059-29-0
302.51
ZPNWNLNPDRYULE-UHFFFAOYSA-N
2-iodobenzene-1-sulfonyl chloride
C6H4ClIO2S
MFCD00833336
627400
ClS(=O)(=O)C1=CC=CC=C1I

Specifications

63059-29-0
135°C
MFCD00833336
3261
ClS(=O)(=O)C1=CC=CC=C1I
302.51
302.51
Crystalline Powder
52°C
C6H4ClIO2S
1 g
ZPNWNLNPDRYULE-UHFFFAOYSA-N
2-iodobenzene-1-sulfonyl chloride
627400
≥98.0% (GC,T)
2-Iodobenzenesulfonyl Chloride
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'