missing translation for 'onlineSavingsMsg'
Learn More

2'-Iodoacetophenone 98.0+%, TCI America™

Catalog No. I06895G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I06895G 5 g
I06891G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I06895G Supplier TCI America Supplier No. I06895G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2142-70-3
Molecular Formula C8H7IO
Molecular Weight (g/mol) 246.047
MDL Number MFCD00094998
InChI Key XDXCBCXNCQGZPG-UHFFFAOYSA-N
Synonym 2'-iodoacetophenone, 1-2-iodophenyl ethanone, o-iodoacetophenone, 1-2-iodophenyl ethan-1-one, o-iodobenzoyl, acetophenone, o-iodo, acetophenone, 2'-iodo, ethanone, 1-iodophenyl, ethanone, 1-2-iodophenyl, 1-acetyl-2-iodobenzene
PubChem CID 240431
IUPAC Name 1-(2-iodophenyl)ethanone
SMILES CC(=O)C1=CC=CC=C1I

Specifications

Color Yellow
Boiling Point 144°C
Quantity 5 g
Formula Weight 246.05
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2′-Iodoacetophenone