missing translation for 'onlineSavingsMsg'
Learn More

2'-Iodoacetophenone, 98+%

Catalog No. AAB2534506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2534506 5 g
AAB2534503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2534506 Supplier Thermo Scientific Chemicals Supplier No. B2534506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 2142-70-3 | C8H7IO | 246.047 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2142-70-3
Molecular Formula C8H7IO
Molecular Weight (g/mol) 246.047
MDL Number MFCD00094998
InChI Key XDXCBCXNCQGZPG-UHFFFAOYSA-N
Synonym 2'-iodoacetophenone, 1-2-iodophenyl ethanone, o-iodoacetophenone, 1-2-iodophenyl ethan-1-one, o-iodobenzoyl, acetophenone, o-iodo, acetophenone, 2'-iodo, ethanone, 1-iodophenyl, ethanone, 1-2-iodophenyl, 1-acetyl-2-iodobenzene
PubChem CID 240431
IUPAC Name 1-(2-iodophenyl)ethanone
SMILES CC(=O)C1=CC=CC=C1I

Specifications

Density 1.746
Boiling Point 139°C to 140°C (12 mmHg)
Flash Point 103°C (218°F)
Refractive Index 1.618
Quantity 5 g
Beilstein 2325078
Sensitivity Light sensitive
Formula Weight 246.05
Percent Purity ≥98%
Chemical Name or Material 2'-Iodoacetophenone
Hazard Category H302+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only