missing translation for 'onlineSavingsMsg'
Learn More

2-Iodoacetamide 98.0+%, TCI America™

Catalog No. p-7131175
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I004425G 25 g
I0044100G 100 g
I0044500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I004425G Supplier TCI America Supplier No. I004425G
Only null left

Chemical Identifiers

CAS 144-48-9
Molecular Formula C2H4INO
Molecular Weight (g/mol) 184.964
MDL Number MFCD00008028
InChI Key PGLTVOMIXTUURA-UHFFFAOYSA-N
Synonym iodoacetamide, monoiodoacetamide, surauto, acetamide, 2-iodo, 2-iodo-acetamide, usaf d-1, alpha-iodoacetamide, ccris 7710, unii-zrh8m27s79, n-3-methoxy-4-butoxybenzyl thiobarbituric acid
PubChem CID 3727
IUPAC Name 2-iodoacetamide
SMILES C(C(=O)N)I

Specifications

Melting Point 95°C
Color White-Yellow
Quantity 25 g
UN Number 2811
Formula Weight 184.96
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 2-Iodoacetamide
EINECSNumber (2)-1152
RTECSNumber AC4200000
TSCA Yes