Learn More
2-Iodoacetamide, 98%, stab. with ca 5-8% water
Description
Alkylating agent used in peptide mapping because it covalently binds with the thiols in cysteine to prevent disulfide bond formation2-Iodoacetamide is used as an electrophile for covalent modification of nucleophilic residues on proteins such as cysteine, methionine and histidine. It is used to bind with thiol group of cysteine, thereby it protects the formation disulfide bonds. It is involved as an inhibitor of deubiquitinase enzymes (DUBs). Further, it acts as an alkylating sulfhydryl reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylating agent used in peptide mapping because it covalently binds with the thiols in cysteine to prevent disulfide bond formation2-Iodoacetamide is used as an electrophile for covalent modification of nucleophilic residues on proteins such as cysteine, methionine and histidine. It is used to bind with thiol group of cysteine, thereby it protects the formation disulfide bonds. It is involved as an inhibitor of deubiquitinase enzymes (DUBs). Further, it acts as an alkylating sulfhydryl reagent.
Solubility
Soluble in water, dimethyl formamide and ethanol. Slightly soluble in methanol.
Notes
Store in a cool place. Light and moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong acids, strong bases, strong oxidizing agents and strong reducing agents.
Specifications
Specifications
| Melting Point | 92°C to 95°(dried) |
| Assay Percent Range | 98%, stabilized with ca 5% water |
| Quantity | 5 g |
| UN Number | UN2811 |
| Beilstein | 1739080 |
| Sensitivity | Light sensitive |
| Solubility Information | Soluble in water,dimethyl formamide and ethanol. Slightly soluble in methanol. |
| Formula Weight | 184.96 |
| Percent Purity | 98% |
| Chemical Name or Material | Iodoacetamide, stabilized with ≈5 to 8% water |
Safety and Handling
| Hazard Category | H301-H314-H317-H334-H335 |
| Hazard Statement | GHS H Statement H301-H315-H319-H317-H335 |
| Precautionary Statement | P260-P264b-P270-P271-P272-P280-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S. |
| EINECSNumber | 205-630-1 |
| RTECSNumber | AC4200000 |
| TSCA | Yes |
| Recommended Storage | Keep cold |
RUO – Research Use Only