missing translation for 'onlineSavingsMsg'
Learn More

2-Iodoacetamide, 98%, stab. with ca 5-8% water

Catalog No. AAA1471522
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1471522 100 g
AAA1471506 5 g
AAA1471514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1471522 Supplier Thermo Scientific Chemicals Supplier No. A1471522
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 144-48-9 | C2H4INO | 184.964 g/mol

Alkylating agent used in peptide mapping because it covalently binds with the thiols in cysteine to prevent disulfide bond formation2-Iodoacetamide is used as an electrophile for covalent modification of nucleophilic residues on proteins such as cysteine, methionine and histidine. It is used to bind with thiol group of cysteine, thereby it protects the formation disulfide bonds. It is involved as an inhibitor of deubiquitinase enzymes (DUBs). Further, it acts as an alkylating sulfhydryl reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Alkylating agent used in peptide mapping because it covalently binds with the thiols in cysteine to prevent disulfide bond formation2-Iodoacetamide is used as an electrophile for covalent modification of nucleophilic residues on proteins such as cysteine, methionine and histidine. It is used to bind with thiol group of cysteine, thereby it protects the formation disulfide bonds. It is involved as an inhibitor of deubiquitinase enzymes (DUBs). Further, it acts as an alkylating sulfhydryl reagent.

Solubility
Soluble in water, dimethyl formamide and ethanol. Slightly soluble in methanol.

Notes
Store in a cool place. Light and moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong acids, strong bases, strong oxidizing agents and strong reducing agents.

Chemical Identifiers

CAS 144-48-9
Molecular Formula C2H4INO
Molecular Weight (g/mol) 184.964
MDL Number MFCD00008028
InChI Key PGLTVOMIXTUURA-UHFFFAOYSA-N
Synonym iodoacetamide, monoiodoacetamide, surauto, acetamide, 2-iodo, 2-iodo-acetamide, usaf d-1, alpha-iodoacetamide, ccris 7710, unii-zrh8m27s79, n-3-methoxy-4-butoxybenzyl thiobarbituric acid
PubChem CID 3727
IUPAC Name 2-iodoacetamide
SMILES C(C(=O)N)I

Specifications

Melting Point 92°C to 95°C (dried)
Odor Characteristic
Assay Percent Range 98%, stabilized with ca 5% water
Quantity 100 g
UN Number UN2811
Beilstein 1739080
Sensitivity Light sensitive
Solubility Information Soluble in water,dimethyl formamide and ethanol. Slightly soluble in methanol.
Formula Weight 184.96
Percent Purity 98%
Chemical Name or Material 2-Iodoacetamide, stabilized with ≈5 to 8% water
Show More Show Less
Hazard Category H301-H314-H317-H334-H335
Hazard Statement GHS H Statement
H301-H315-H319-H317-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P270-P271-P272-P280-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 205-630-1
RTECSNumber AC4200000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only