missing translation for 'onlineSavingsMsg'
Learn More

2-Iodo-5-nitropyridine 98.0+%, TCI America™

Catalog No. I09551G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I09551G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I09551G Supplier TCI America Supplier No. I09551G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 28080-54-8
Molecular Formula C5H3IN2O2
Molecular Weight (g/mol) 249.995
MDL Number MFCD05863241
InChI Key SJXWHBQWFBHASX-UHFFFAOYSA-N
Synonym 6-iodo-3-nitropyridine, acmc-20alyk, pubchem6575, 5-nitro-2-iodopyridine, pyridine,2-iodo-5-nitro, 2-iodanyl-5-nitro-pyridine, pyridine, 2-iodo-5-nitro, 2-iodo-5-nitropyridine
PubChem CID 819492
IUPAC Name 2-iodo-5-nitropyridine
SMILES C1=CC(=NC=C1[N+](=O)[O-])I

Specifications

Melting Point 166°C
Color White-Yellow
Quantity 1 g
Formula Weight 250.00
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Iodo-5-nitropyridine