missing translation for 'onlineSavingsMsg'
Learn More

2-Iodo-5-nitropyridine, 97%

Catalog No. AAH2710306
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2710306 5 g
AAH2710303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2710306 Supplier Thermo Scientific Chemicals Supplier No. H2710306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 28080-54-8 | C5H3IN2O2 | 249.995 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 28080-54-8
Molecular Formula C5H3IN2O2
Molecular Weight (g/mol) 249.995
MDL Number MFCD05863241
InChI Key SJXWHBQWFBHASX-UHFFFAOYSA-N
PubChem CID 819492
IUPAC Name 2-iodo-5-nitropyridine
SMILES C1=CC(=NC=C1[N+](=O)[O-])I

Specifications

Melting Point 163°C to 168°C
Quantity 5 g
Sensitivity Light sensitive
Synonym 6-iodo-3-nitropyridine, acmc-20alyk, pubchem6575, 5-nitro-2-iodopyridine, pyridine,2-iodo-5-nitro, 2-iodanyl-5-nitro-pyridine, pyridine, 2-iodo-5-nitro, 2-iodo-5-nitropyridine
Formula Weight 249.98
Percent Purity 97%
Chemical Name or Material 2-Iodo-5-nitropyridine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only