missing translation for 'onlineSavingsMsg'
Learn More

2-Iodo-3-methoxypyridine, 97%

Catalog No. AAL1986806
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1986806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1986806 Supplier Thermo Scientific Chemicals Supplier No. L1986806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 93560-55-5 | C6H6INO | 235.024 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 93560-55-5
Molecular Formula C6H6INO
Molecular Weight (g/mol) 235.024
MDL Number MFCD00234181
InChI Key NJFRZBAZMPWJKQ-UHFFFAOYSA-N
Synonym 2-iodanyl-3-methoxy-pyridine, pyridine, 2-iodo-3-methoxy, 2-iodo-3-methoxy-pyridine
PubChem CID 817173
IUPAC Name 2-iodo-3-methoxypyridine
SMILES COC1=C(N=CC=C1)I

Specifications

Melting Point 57°C to 61°C
Quantity 5 g
Beilstein 115569
Sensitivity Light sensitive
Formula Weight 235.03
Percent Purity 97%
Chemical Name or Material 2-Iodo-3-methoxypyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only