missing translation for 'onlineSavingsMsg'
Learn More

2-Iodo-2-methylpropane (stabilized with Copper chip) 97.0+%, TCI America™

Catalog No. B107225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B107225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B107225G Supplier TCI America Supplier No. B107225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 558-17-8
Molecular Formula C4H9I
Molecular Weight (g/mol) 184.02
MDL Number MFCD00001068
InChI Key ANGGPYSFTXVERY-UHFFFAOYSA-N
Synonym tert-butyl iodide, propane, 2-iodo-2-methyl, trimethyliodomethane, t-butyl iodide, unii-e5hy19g10p, 2-jod-2-methylpropan, sjphavimup@, tert-c4h9i, 2-iodo-2-methyl propane, 2-iodo-2-methyl-propane
PubChem CID 11206
IUPAC Name 2-iodo-2-methylpropane
SMILES CC(C)(C)I

Specifications

Melting Point -38°C
Color Yellow
Boiling Point 99°C
Quantity 25 g
UN Number 2391
Formula Weight 184.02
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Iodo-2-methylpropane (stabilized with Copper chip)
RTECSNumber TZ4251000
TSCA Yes
Recommended Storage Refrigerator