missing translation for 'onlineSavingsMsg'
Learn More

2-Iodo-2-methylpropane, 95%, stab. with copper powder

Catalog No. p-7043799
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0278614 25 g
AAL0278622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0278614 Supplier Thermo Scientific Chemicals Supplier No. L0278614
Only null left

CAS: 558-17-8 | C4H9I | 184.02 g/mol

2-Iodo-2-methylpropane is used as a chemical reagent in organic synthesis. Further, it serves as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Iodo-2-methylpropane is used as a chemical reagent in organic synthesis. Further, it serves as a pharmaceutical intermediate.

Solubility
Miscible with ethanol and ether.

Notes
Light sensitive. Incompatible with strong oxidizing agents and strong bases.

Chemical Identifiers

CAS 558-17-8
Molecular Formula C4H9I
Molecular Weight (g/mol) 184.02
MDL Number MFCD00001068
InChI Key ANGGPYSFTXVERY-UHFFFAOYSA-N
Synonym tert-butyl iodide, propane, 2-iodo-2-methyl, trimethyliodomethane, t-butyl iodide, unii-e5hy19g10p, 2-jod-2-methylpropan, sjphavimup@, tert-c4h9i, 2-iodo-2-methyl propane, 2-iodo-2-methyl-propane
PubChem CID 11206
IUPAC Name 2-iodo-2-methylpropane
SMILES CC(C)(C)I

Specifications

Melting Point -38°C
Density 1.547
Boiling Point 99°C to 100°C
Flash Point 7°C (44°F)
Odor Recognizable
Refractive Index 1.491
Quantity 25 g
UN Number UN2391
Beilstein 505950
Sensitivity Light sensitive
Solubility Information Miscible with ethanol and ether.
Formula Weight 184.02
Percent Purity 95%
Chemical Name or Material 2-Iodo-2-methylpropane, stabilized with copper powder
Show More Show Less
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: IODOMETHYLPROPANES
EINECSNumber 209-190-1
RTECSNumber TZ4251000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only