missing translation for 'onlineSavingsMsg'
Learn More

2-Iodo-1-methylimidazole, 97%

Catalog No. AAH51862MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH51862MD 250 mg
AAH5186203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH51862MD Supplier Thermo Scientific Chemicals Supplier No. H51862MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 37067-95-1 | C4H5IN2 | 208.002 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 37067-95-1
Molecular Formula C4H5IN2
Molecular Weight (g/mol) 208.002
MDL Number MFCD00661606
InChI Key BKRACZNEJSCZML-UHFFFAOYSA-N
Synonym 2-iodo-1-methyl-1h-imidazole, 1h-imidazole, 2-iodo-1-methyl, pubchem16310, acmc-1ajih, 2-iodo-1-methyl-imidazole, 2-iodanyl-1-methyl-imidazole, 1-methyl-2-iodo-1h-imidazole, 2-iodo-n-methylimidazole, 1-methyl-2-iodoimidazole, 1h-imidazole,2-iodo-1-methyl
PubChem CID 335835
IUPAC Name 2-iodo-1-methylimidazole
SMILES CN1C=CN=C1I

Specifications

Melting Point 79°C to 82°C
Quantity 250 mg
Sensitivity Light sensitive
Formula Weight 208
Percent Purity 97%
Chemical Name or Material 2-Iodo-1-methylimidazole
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only