missing translation for 'onlineSavingsMsg'
Learn More

2-Iodo-1-(2-tetrahydropyranyl)-1H-imidazole, 95%, Thermo Scientific™

Catalog No. p-7030945
Change view
Click to view available options
Quantity:
250mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH31846MD 250mg
AAH3184603 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH31846MD Supplier Thermo Scientific Chemicals Supplier No. H31846MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 1047626-77-6
Molecular Formula C8H11IN2O
Molecular Weight (g/mol) 278.093
MDL Number MFCD11100955
InChI Key UWFGOSIGMHYERY-UHFFFAOYSA-N
Synonym 2-iodo-1-tetrahydro-2h-pyran-2-yl-1h-imidazole, 2-iodo-1-2-tetrahydropyranyl-1h-imidazole, 2-iodo-1-oxan-2-yl imidazole, 2-iodo-1-oxan-2-yl-1h-imidazole
PubChem CID 45588154
IUPAC Name 2-iodo-1-(oxan-2-yl)imidazole
SMILES C1CCOC(C1)N2C=CN=C2I

Specifications

Quantity 250mg
Sensitivity Light sensitive
Formula Weight 278.09
Percent Purity 95%
Chemical Name or Material 2-Iodo-1-(2-tetrahydropyranyl)-1H-imidazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only