missing translation for 'onlineSavingsMsg'
Learn More

2-Iodo-1,1,1-trifluoroethane, 99%

Catalog No. AC176080500
Change view
Click to view available options
Quantity:
10 mL
50 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC176080500 50 mL Glass bottle
AC176080100 10 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC176080500 Supplier Thermo Scientific Chemicals Supplier No. 176080500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 353-83-3 | C2H2F3I | 209.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 353-83-3
Molecular Formula C2H2F3I
Molecular Weight (g/mol) 209.94
MDL Number MFCD00001082
InChI Key RKOUFQLNMRAACI-UHFFFAOYSA-N
Synonym 2-iodo-1,1,1-trifluoroethane, 2,2,2-trifluoroethyl iodide, trifluoroethyl iodide, ethane, 1,1,1-trifluoro-2-iodo, 1-iodo-2,2,2-trifluoroethane, 2,2,2-trifluoroiodoethane, cf3ch2i, unii-ca0j38we9z, 1,1,1-trifluoroiodoethane, 1,1,1-trifluoro-2-iodo-ethane
PubChem CID 67709
IUPAC Name 1,1,1-trifluoro-2-iodoethane
SMILES C(C(F)(F)F)I

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Pink
Density 2.1300g/mL
Boiling Point 53.0°C to 55.0°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4000 to 1.4020
Linear Formula ICH2CF3
Quantity 50 mL
Specific Gravity 2.13
Solubility Information Solubility in water: insoluble
Formula Weight 209.94
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Iodo-1, 1, 1-trifluoroethane, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 206-541-0
TSCA TSCA

RUO – Research Use Only