missing translation for 'onlineSavingsMsg'
Learn More

2-Indanone, 97%

Catalog No. AC164110100
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC164110100 10 g Glass bottle
AC164110500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC164110100 Supplier Thermo Scientific Chemicals Supplier No. 164110100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 615-13-4 | C9H8O | 132.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 615-13-4
Molecular Formula C9H8O
Molecular Weight (g/mol) 132.16
MDL Number MFCD00003792
InChI Key UMJJFEIKYGFCAT-UHFFFAOYSA-N
Synonym 2-indanone, indan-2-one, 1h-inden-2 3h-one, 1,3-dihydro-2h-inden-2-one, beta-hydrindone, 2h-inden-2-one, 1,3-dihydro, unii-0i79n673de, 2,3-dihydro-1h-inden-2-one, 4-07-00-01002 beilstein handbook reference, 2-lndanone
PubChem CID 11983
ChEBI CHEBI:27930
IUPAC Name 1,3-dihydroinden-2-one
SMILES C1C(=O)CC2=CC=CC=C21

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 50.0°C to 57.0°C
Color Brown-Yellow to Yellow
Flash Point 100°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 10 g
Beilstein 07, 363
Solubility Information Solubility in water: insoluble
Formula Weight 132.16
Percent Purity 97%
Physical Form Crystals or Powder
Chemical Name or Material 2-Indanone, 97%
Show More Show Less
EINECSNumber 210-410-3
RTECSNumber NK7535500
TSCA TSCA

RUO – Research Use Only