missing translation for 'onlineSavingsMsg'
Learn More

2-Iminothiolane Hydrochloride, 98%

Catalog No. p-4812814
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC450182500 250 mg
AC450180010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC450182500 Supplier Thermo Scientific Chemicals Supplier No. 450182500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4781-83-3 | C4H7NS·ClH | 137.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4781-83-3
Molecular Formula C4H7NS·ClH
Molecular Weight (g/mol) 137.63
InChI Key ATGUDZODTABURZ-UHFFFAOYSA-N
Synonym 2-iminothiolane hydrochloride, 2-iminothiolane hcl, thiolan-2-imine hydrochloride, dihydrothiophen-2 3h-imine hydrochloride, 2 3h-thiophenimine, dihydro-, hydrochloride, traut's reagent, c4h7ns.clh, traut eo 1/4 a, 2-thiolanimine hydrochloride, 2 3h-thiophenimine, hydrochloride
PubChem CID 13166855
IUPAC Name thiolan-2-imine;hydrochloride
SMILES C1CC(=N)SC1.Cl

Specifications

Assay Percent Range 97.5% min. (Argentometry)
Packaging Glass Bottle
Quantity 250 mg
Formula Weight 137.63
Percent Purity 98%
Chemical Name or Material 2-Iminothiolane Hydrochloride
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only