missing translation for 'onlineSavingsMsg'
Learn More

2-Imidazolecarboxylic Acid 98.0+%, TCI America™

Catalog No. p-7131712
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
I10081G 1 g
I10085G 5 g
2 options

Catalog No. I10081G

Supplier: TCI America I10081G

Only null left

Chemical Identifiers

CAS 16042-25-4
Molecular Formula C4H4N2O2
Molecular Weight (g/mol) 112.088
MDL Number MFCD00270816
InChI Key KYWMCFOWDYFYLV-UHFFFAOYSA-N
Synonym 2-imidazolecarboxylic acid, imidazole-2-carboxylic acid, unii-o9g6515vc1, 1h-imidazolecarboxylic acid, 2-imidazolecarboxylicacid, 2-imidazocarboxylic acid, 1h-imidazole-2-carboxylicacid, imidazolecarboxylic, acmc-1avrt, 2-carboxy-1h-imidazole
PubChem CID 574321
IUPAC Name 1H-imidazole-2-carboxylic acid
SMILES C1=CN=C(N1)C(=O)O

Specifications

Melting Point 164°C
Color Blue-White
Quantity 1 g
Formula Weight 112.09
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Imidazolecarboxylic Acid