missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxysaclofen, Thermo Scientific Chemicals

Selective antagonist at GABA-B receptors

$889.71 - $889.71

Chemical Identifiers

CAS 117354-64-0
Molecular Formula C9H12ClNO4S
Molecular Weight (g/mol) 265.708
MDL Number MFCD00069282
InChI Key WBSMZVIMANOCNX-UHFFFAOYSA-N
Synonym 2-hydroxysaclofen, 2-oh-saclofen, 2-hydroxy-saclofen, 3-amino-2-4-chlorophenyl-2-hydroxypropane-1-sulfonic acid, 2-hydroxy-s---saclofen, 3-amino-2-4-chlorophenyl-2-hydroxypropylsulfonic acid, 3-amino-2-4-chlorophenyl-2-hydroxypropanesulfonic acid, beta-aminomethyl-4-chloro-beta-hydroxybenzeneethanesulfonic acid, benzeneethanesulfonic acid, beta-aminomethyl-4-chloro-beta-hydroxy, 2-hydroxysaclofen, solid
PubChem CID 1564
IUPAC Name 3-amino-2-(4-chlorophenyl)-2-hydroxypropane-1-sulfonic acid
SMILES C1=CC(=CC=C1C(CN)(CS(=O)(=O)O)O)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ63588MA
View Documents
Thermo Scientific Chemicals
J63588MA
10 mg
Each for $889.71
Only null left
 
Description

Description

Selective antagonist at GABA-B receptors

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

117354-64-0
265.708
WBSMZVIMANOCNX-UHFFFAOYSA-N
1564
C1=CC(=CC=C1C(CN)(CS(=O)(=O)O)O)Cl
C9H12ClNO4S
MFCD00069282
2-hydroxysaclofen, 2-oh-saclofen, 2-hydroxy-saclofen, 3-amino-2-4-chlorophenyl-2-hydroxypropane-1-sulfonic acid, 2-hydroxy-s---saclofen, 3-amino-2-4-chlorophenyl-2-hydroxypropylsulfonic acid, 3-amino-2-4-chlorophenyl-2-hydroxypropanesulfonic acid, beta-aminomethyl-4-chloro-beta-hydroxybenzeneethanesulfonic acid, benzeneethanesulfonic acid, beta-aminomethyl-4-chloro-beta-hydroxy, 2-hydroxysaclofen, solid
3-amino-2-(4-chlorophenyl)-2-hydroxypropane-1-sulfonic acid

Specifications

268°C to 271°C
White
Solid
MFCD00069282
2-hydroxysaclofen, 2-oh-saclofen, 2-hydroxy-saclofen, 3-amino-2-4-chlorophenyl-2-hydroxypropane-1-sulfonic acid, 2-hydroxy-s---saclofen, 3-amino-2-4-chlorophenyl-2-hydroxypropylsulfonic acid, 3-amino-2-4-chlorophenyl-2-hydroxypropanesulfonic acid, beta-aminomethyl-4-chloro-beta-hydroxybenzeneethanesulfonic acid, benzeneethanesulfonic acid, beta-aminomethyl-4-chloro-beta-hydroxy, 2-hydroxysaclofen, solid
WBSMZVIMANOCNX-UHFFFAOYSA-N
3-amino-2-(4-chlorophenyl)-2-hydroxypropane-1-sulfonic acid
1564
2-Hydroxysaclofen
117354-64-0
10 mg
C9H12ClNO4S
UN1759
Soluble in water at 1mg/ml; Also soluble in DMSO,0;1M HCl and methanol
C1=CC(=CC=C1C(CN)(CS(=O)(=O)O)O)Cl
265.708
265.71
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.

RTECSNumber : DA0359200

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only