missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxyquinoxaline 98.0+%, TCI America™

Catalog No. H0591500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0591500G 500 g
H059125G 25 g
H0591100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H0591500G Supplier TCI America Supplier No. H0591500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1196-57-2
Molecular Formula C8H6N2O
Molecular Weight (g/mol) 146.15
MDL Number MFCD00006722
InChI Key FFRYUAVNPBUEIC-UHFFFAOYSA-N
Synonym 2-hydroxyquinoxaline, quinoxalin-2-ol, 2-quinoxalinol, 2 1h-quinoxalinone, quinoxalin-2 1h-one, quinoxalin-2-one, 2-quinoxalinone, 3-quinoxalinone, 2-quinoxalone, 1,2-dihydroquinoxalin-2-one
PubChem CID 14526
ChEBI CHEBI:38890
IUPAC Name 1,2-dihydroquinoxalin-2-one
SMILES O=C1NC2=CC=CC=C2N=C1

Specifications

Melting Point 275°C
Quantity 500 g
Formula Weight 146.15
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxyquinoxaline
EINECSNumber (5)-6201
RTECSNumber VD3630000
TSCA Yes