missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxypyrimidine Hydrochloride 98.0+%, TCI America™

Catalog No. p-7130558
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H048025G 25 g
H0480250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H048025G Supplier TCI America Supplier No. H048025G
Only null left

Chemical Identifiers

CAS 38353-09-2
Molecular Formula C4H5ClN2O
Molecular Weight (g/mol) 132.547
MDL Number MFCD00012781
InChI Key IAJINJSFYTZPEJ-UHFFFAOYSA-N
Synonym 2-hydroxypyrimidine hydrochloride, pyrimidin-2-ol hydrochloride, 2 1h-pyrimidinone, monohydrochloride, 2-hydroxypyrimidine hcl, 2-pyrimidinone hydrochloride, pyrimidin-2 1h-one hydrochloride, 2-hydroxypyrimidinehydrochloride, pyrimidin-2-one monohydrochloride, 1,2-dihydropyrimidin-2-one hydrochloride, pyrimidin-2-one hydrochloride
PubChem CID 122774
IUPAC Name 1H-pyrimidin-2-one;hydrochloride
SMILES C1=CNC(=O)N=C1.Cl

Specifications

Quantity 25 g
Formula Weight 132.55
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxypyrimidine Hydrochloride