missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxypyrimidine-5-boronic acid pinacol ester, 95%

Catalog No. aah2697603
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
250 mg
Catalog No. Quantity
AAH2697603 1 g
AAH26976MD 250 mg
2 options

Catalog No. AAH2697603

Supplier: Thermo Scientific Chemicals H2697603

Only null left

CAS: 1073354-84-3 | C10H15BN2O3 | 222.051 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1073354-84-3
Molecular Formula C10H15BN2O3
Molecular Weight (g/mol) 222.051
MDL Number MFCD09037497
InChI Key JRCSRFGCQSLWNZ-UHFFFAOYSA-N
Synonym 2-hydroxypyrimidine-5-boronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidin-2-ol, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyrimidin-2-ol, 5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidin-2-ol, 2-hydroxypyrimidin-5-ylboronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-pyrimidinol, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidin-2 1h-one
PubChem CID 44755196
IUPAC Name 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrimidin-2-one
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CNC(=O)N=C2

Specifications

Melting Point 201°C to 204°C
Quantity 1 g
Formula Weight 222.05
Percent Purity 95%
Chemical Name or Material 2-Hydroxypyrimidine-5-boronic acid pinacol ester
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only