missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxypalmitic Acid 97.0+%, TCI America™

Catalog No. H04031G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H04031G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H04031G Supplier TCI America Supplier No. H04031G
Only null left

Chemical Identifiers

CAS 764-67-0
Molecular Formula C16H32O3
Molecular Weight (g/mol) 272.43
MDL Number MFCD00014343
InChI Key JGHSBPIZNUXPLA-UHFFFAOYNA-N
Synonym 2-Hydroxyhexadecanoic Acid
PubChem CID 92836
ChEBI CHEBI:65101
IUPAC Name 2-hydroxyhexadecanoic acid
SMILES CCCCCCCCCCCCCCC(O)C(O)=O

Specifications

Melting Point 86°C
Color White
Quantity 1 g
Formula Weight 272.43
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxypalmitic Acid
EINECSNumber (2)-1340
TSCA No