missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxynicotinic acid, 98%

Catalog No. p-146552
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC247220250 25 g Glass bottle
AC247221000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC247220250 Supplier Thermo Scientific Chemicals Supplier No. 247220250

CAS: 609-71-2 | C6H5NO3 | 139.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 609-71-2
Molecular Formula C6H5NO3
Molecular Weight (g/mol) 139.11
MDL Number MFCD00010100
InChI Key UEYQJQVBUVAELZ-UHFFFAOYSA-N
Synonym 2-hydroxynicotinic acid, 2-hydroxypyridine-3-carboxylic acid, 2-oxo-1,2-dihydropyridine-3-carboxylic acid, 2-hydroxynicotinicacid, unii-tmq68q9ba3, 3-pyridinecarboxylic acid, 1,2-dihydro-2-oxo, 1,2-dihydro-2-oxonicotinic acid, tmq68q9ba3, 2-hydroxy-3-pyridinecarboxylic acid, 1,2-dihydro-2-oxo-3-pyridinecarboxylic acid
PubChem CID 69114
IUPAC Name 2-oxo-1H-pyridine-3-carboxylic acid
SMILES OC(=O)C1=CC=CNC1=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 259.0°C to 261.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 25 g
Beilstein 22, 214
Merck Index 10, 2822
Formula Weight 139.11
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Hydroxynicotinic acid, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 210-198-2
TSCA TSCA

RUO – Research Use Only