missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxymethyloxetane 95.0+%, TCI America™

Catalog No. H07731G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H07731G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H07731G Supplier TCI America Supplier No. H07731G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 61266-70-4
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00142933
InChI Key PQZJTHGEFIQMCO-UHFFFAOYSA-N
Synonym 2-hydroxymethyloxetane, 2-oxetanemethanol, 2-oxetanylmethanol, 2,4-epoxybutanol, oxetan-2-yl-methanol, oxetane-2-methanol, methyloloxacyclobutan, 2-hydrpxymethyloxetane, oxetan-2-yl methanol, rac-2-hydroxymethyloxetane
PubChem CID 12284142
IUPAC Name oxetan-2-ylmethanol
SMILES C1COC1CO

Specifications

Color Yellow
Boiling Point 98°C
Quantity 1 g
Formula Weight 88.11
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Hydroxymethyloxetane
TSCA No
Recommended Storage Refrigerator