missing translation for 'onlineSavingsMsg'
Learn More

2-(Hydroxymethyl)pyridine, 98%

Catalog No. p-145940
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC131990250 25 g Glass bottle
AC131991000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC131990250 Supplier Thermo Scientific Chemicals Supplier No. 131990250
Only null left

CAS: 586-98-1 | C6H7NO | 109.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 586-98-1
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.13
MDL Number MFCD00006348
InChI Key SHNUBALDGXWUJI-UHFFFAOYSA-N
Synonym 2-pyridinemethanol, 2-hydroxymethyl pyridine, piconol, 2-pyridinylmethanol, 2-pyridylmethanol, 2-pyridylcarbinol, pyridine-2-methanol, pyridine-2-carbinol, alpha-picolyl alcohol, pyridin-2-yl methanol
PubChem CID 11474
IUPAC Name pyridin-2-ylmethanol
SMILES C1=CC=NC(=C1)CO

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 5.0°C
Color Brown or Colorless to Yellow
Density 1.1300g/mL
Boiling Point 220.0°C
Flash Point 117°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5410 to 1.5440
Linear Formula (C5H4N)CH2OH
Quantity 25 g
Beilstein 21, I, 203
Specific Gravity 1.13
Solubility Information Solubility in water: soluble
Formula Weight 109.13
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-(Hydroxymethyl)pyridine, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye pr
EINECSNumber 209-592-7
RTECSNumber UT4689000
TSCA TSCA

RUO – Research Use Only