missing translation for 'onlineSavingsMsg'
Learn More

2-(Hydroxymethyl)-18-crown 6-Ether 93.0+%, TCI America™

Catalog No. p-7130847
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0990200MG 200 mg
H09901G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H0990200MG Supplier TCI America Supplier No. H0990200MG
Only null left

Chemical Identifiers

CAS 70069-04-4
Molecular Formula C13H26O7
Molecular Weight (g/mol) 294.34
MDL Number MFCD00188046
InChI Key HFRGASADQCZXHH-UHFFFAOYNA-N
Synonym 1,4,7,10,13,16-hexaoxacyclooctadecane-2-methanol, 2-hydroxymethyl-18-crown-6, 2-hydroxymethyl-18-crown 6-ether, 1,4,7,10,13,16-hexaoxacyclooctadecan-2-ylmethanol, hydroxy-18-crown-6, 18-crown-6-methanol, hydroxymethyl-18-crown-6, hfrgasadqczxhh-uhfffaoysa, 2-hydroxymethyl-18-crown, 2,5,8,11,14,17-hexaoxacyclooctadecyl methan-1-ol
PubChem CID 3611393
IUPAC Name (1,4,7,10,13,16-hexaoxacyclooctadecan-2-yl)methanol
SMILES OCC1COCCOCCOCCOCCOCCO1

Specifications

Quantity 200 mg
Formula Weight 294.34
Percent Purity ≥93.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(Hydroxymethyl)-18-crown 6-Ether