missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Hydroxyméthyl)-18-couronne 6-Éther 93,0+%, TCI America™
Spécifications
Spécifications
| Quantity | 200 mg |
| Formula Weight | 294.34 |
| Percent Purity | ≥93.0% (GC) |
| Physical Form | Liquide |
| Chemical Name or Material | 2-(Hydroxymethyl)-18-crown 6-Ether |
Sécurité et manipulation
| TSCA | No |