missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxymethyl-18-crown-6, 97%

Catalog No. p-7059368
Change view
Click to view available options
Quantity:
0.25 g
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4447701 0.25 g
AA4447703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4447701 Supplier Thermo Scientific Chemicals Supplier No. 04447701

CAS: 70069-04-4 | C13H26O7 | 294.34 g/mol

2-Hydroxymethyl-18-crown-6 is used for chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 70069-04-4
Molecular Formula C13H26O7
Molecular Weight (g/mol) 294.34
MDL Number MFCD00188046
InChI Key HFRGASADQCZXHH-UHFFFAOYNA-N
Synonym 1,4,7,10,13,16-hexaoxacyclooctadecane-2-methanol, 2-hydroxymethyl-18-crown-6, 2-hydroxymethyl-18-crown 6-ether, 1,4,7,10,13,16-hexaoxacyclooctadecan-2-ylmethanol, hydroxy-18-crown-6, 18-crown-6-methanol, hydroxymethyl-18-crown-6, hfrgasadqczxhh-uhfffaoysa, 2-hydroxymethyl-18-crown, 2,5,8,11,14,17-hexaoxacyclooctadecyl methan-1-ol
PubChem CID 3611393
IUPAC Name 1,4,7,10,13,16-hexaoxacyclooctadec-2-ylmethanol
SMILES OCC1COCCOCCOCCOCCOCCO1

Specifications

Density 1.174 g/mL
Boiling Point 150°C (0.5 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.479
Quantity 0.25 g
Beilstein 4313301
Solubility Information Soluble in water (1000g/L) (25°C).
Formula Weight 294.35
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Hydroxymethyl-18-crown-6
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 274-299-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only