missing translation for 'onlineSavingsMsg'
Learn More

2-(Hydroxymethyl)-15-crown 5-Ether 96.0+%, TCI America™

Catalog No. p-7130842
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H09831G 1 g
H09835G 5 g
H098325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H09831G Supplier TCI America Supplier No. H09831G

Chemical Identifiers

CAS 75507-25-4
Molecular Formula C11H22O6
Molecular Weight (g/mol) 250.291
MDL Number MFCD00192215
InChI Key YHIQMMGCRYKJLB-UHFFFAOYSA-N
PubChem CID 2733744
IUPAC Name 1,4,7,10,13-pentaoxacyclopentadec-2-ylmethanol
SMILES C1COCCOCC(OCCOCCO1)CO

Specifications

Color Yellow
Quantity 1 g
Synonym 1,4,7,10,13-pentaoxacyclopentadecane-2-methanol, 2-hydroxymethyl-15-crown-5, 2-hydroxymethyl-15-crown 5-ether, 1,4,7,10,13-pentaoxacyclopentadecan-2-ylmethanol, hydroxy-15-crown-5, 2-hydroxymethyl-15-crown5-ether, 2-hydroxymethyl-15-crown, 2,5,8,11,14-pentaoxacyclopentadecyl methan-1-ol, 1,4,7,10,13-pentaoxacyclopentadec-2-yl-methanol, 1,4,7,10,13-pentaoxacyclopentadecan-2-yl methanol
Formula Weight 250.29
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(Hydroxymethyl)-15-crown 5-Ether