missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Hydroxymethyl)-12-crown 4-Ether 95.0+%, TCI America™
$1172.49 - $4546.43
Chemical Identifiers
| CAS | 75507-26-5 |
|---|---|
| Molecular Formula | C9H18O5 |
| Molecular Weight (g/mol) | 206.238 |
| MDL Number | MFCD00067524 |
| InChI Key | NJIPEIQHUNDGPY-UHFFFAOYSA-N |
| Synonym | 1,4,7,10-tetraoxacyclododecane-2-methanol, 2-hydroxymethyl-12-crown-4, 2-hydroxymethyl-12-crown4-ether, 1,4,7,10-tetraoxacyclododecan-2-ylmethanol, 2-hydroxymethyl-12-crown, 2-hydroxymethyl-12-crown 4-ether, 2-hydroxymethyl-12-crown-4-ether, 2-hydroxymethyl-1,2-crown 4-ether, 1,4,7,10-tetraoxacyclododecan-2-methanol, 2,5,8,11-tetraoxacyclododecyl methan-1-ol |
| PubChem CID | 2733085 |
| IUPAC Name | 1,4,7,10-tetraoxacyclododec-2-ylmethanol |
| SMILES | C1COCCOC(COCCO1)CO |
Chemical Identifiers
| 75507-26-5 | |
| 206.238 | |
| NJIPEIQHUNDGPY-UHFFFAOYSA-N | |
| 2733085 | |
| C1COCCOC(COCCO1)CO |
| C9H18O5 | |
| MFCD00067524 | |
| 1,4,7,10-tetraoxacyclododecane-2-methanol, 2-hydroxymethyl-12-crown-4, 2-hydroxymethyl-12-crown4-ether, 1,4,7,10-tetraoxacyclododecan-2-ylmethanol, 2-hydroxymethyl-12-crown, 2-hydroxymethyl-12-crown 4-ether, 2-hydroxymethyl-12-crown-4-ether, 2-hydroxymethyl-1,2-crown 4-ether, 1,4,7,10-tetraoxacyclododecan-2-methanol, 2,5,8,11-tetraoxacyclododecyl methan-1-ol | |
| 1,4,7,10-tetraoxacyclododec-2-ylmethanol |
Specifications
| 75507-26-5 | |
| C9H18O5 | |
| 1 g | |
| NJIPEIQHUNDGPY-UHFFFAOYSA-N | |
| 1,4,7,10-tetraoxacyclododec-2-ylmethanol | |
| 2733085 | |
| ≥95.0% (GC) | |
| 2-(Hydroxymethyl)-12-crown 4-Ether |
| Yellow | |
| MFCD00067524 | |
| 1,4,7,10-tetraoxacyclododecane-2-methanol, 2-hydroxymethyl-12-crown-4, 2-hydroxymethyl-12-crown4-ether, 1,4,7,10-tetraoxacyclododecan-2-ylmethanol, 2-hydroxymethyl-12-crown, 2-hydroxymethyl-12-crown 4-ether, 2-hydroxymethyl-12-crown-4-ether, 2-hydroxymethyl-1,2-crown 4-ether, 1,4,7,10-tetraoxacyclododecan-2-methanol, 2,5,8,11-tetraoxacyclododecyl methan-1-ol | |
| C1COCCOC(COCCO1)CO | |
| 206.238 | |
| 206.24 | |
| Liquid |
Safety and Handling
TSCA : No