missing translation for 'onlineSavingsMsg'
Learn More

2-(Hydroxymethyl)-12-crown 4-Ether 95.0+%, TCI America™

Catalog No. H09821G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H09821G 1 g
H09825G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H09821G Supplier TCI America Supplier No. H09821G

Chemical Identifiers

CAS 75507-26-5
Molecular Formula C9H18O5
Molecular Weight (g/mol) 206.238
MDL Number MFCD00067524
InChI Key NJIPEIQHUNDGPY-UHFFFAOYSA-N
Synonym 1,4,7,10-tetraoxacyclododecane-2-methanol, 2-hydroxymethyl-12-crown-4, 2-hydroxymethyl-12-crown4-ether, 1,4,7,10-tetraoxacyclododecan-2-ylmethanol, 2-hydroxymethyl-12-crown, 2-hydroxymethyl-12-crown 4-ether, 2-hydroxymethyl-12-crown-4-ether, 2-hydroxymethyl-1,2-crown 4-ether, 1,4,7,10-tetraoxacyclododecan-2-methanol, 2,5,8,11-tetraoxacyclododecyl methan-1-ol
PubChem CID 2733085
IUPAC Name 1,4,7,10-tetraoxacyclododec-2-ylmethanol
SMILES C1COCCOC(COCCO1)CO

Specifications

Color Yellow
Quantity 1 g
Formula Weight 206.24
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(Hydroxymethyl)-12-crown 4-Ether