Learn More
2-Hydroxymethyl-12-crown-4, 97%
CAS: 75507-26-5 | C9H18O5 | 206.238 g/mol
$89.67 - $910.90
Chemical Identifiers
| CAS | 75507-26-5 |
|---|---|
| Molecular Formula | C9H18O5 |
| Molecular Weight (g/mol) | 206.238 |
| MDL Number | MFCD00067524 |
| InChI Key | NJIPEIQHUNDGPY-UHFFFAOYSA-N |
| PubChem CID | 2733085 |
| IUPAC Name | 1,4,7,10-tetraoxacyclododec-2-ylmethanol |
| SMILES | C1COCCOC(COCCO1)CO |
Description
Ethylene oxide is important or critical to the production of detergents, thickeners, solvents, plastics, and various organic chemicals such as ethylene glycol, ethanolamines, simple and complex glycols, polyglycol ethers and other compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 75507-26-5 | |
| 206.238 | |
| NJIPEIQHUNDGPY-UHFFFAOYSA-N | |
| 1,4,7,10-tetraoxacyclododec-2-ylmethanol |
| C9H18O5 | |
| MFCD00067524 | |
| 2733085 | |
| C1COCCOC(COCCO1)CO |
Specifications
| 75507-26-5 | |
| 115°C (0.04 mmHg) | |
| C9H18O5 | |
| MFCD00067524 | |
| 4661968 | |
| Miscible in water. Soluble in non-polar solvents. Insoluble in water. | |
| C1COCCOC(COCCO1)CO | |
| 206.238 | |
| 206.24 | |
| Liquid |
| 1.20 g/mL | |
| >110°C (230°F) | |
| 1.48 | |
| 0.25 g | |
| 1,4,7,10-tetraoxacyclododecane-2-methanol, 2-hydroxymethyl-12-crown-4, 2-hydroxymethyl-12-crown4-ether, 1,4,7,10-tetraoxacyclododecan-2-ylmethanol, 2-hydroxymethyl-12-crown, 2-hydroxymethyl-12-crown 4-ether, 2-hydroxymethyl-12-crown-4-ether, 2-hydroxymethyl-1,2-crown 4-ether, 1,4,7,10-tetraoxacyclododecan-2-methanol, 2,5,8,11-tetraoxacyclododecyl methan-1-ol | |
| NJIPEIQHUNDGPY-UHFFFAOYSA-N | |
| 1,4,7,10-tetraoxacyclododec-2-ylmethanol | |
| 2733085 | |
| 97% | |
| 2-Hydroxymethyl-12-crown-4 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 278-224-5
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only