missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxymethyl-1-methylimidazole 98.0+%, TCI America™

Catalog No. H13701G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H13701G 1 g
H13705G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H13701G Supplier TCI America Supplier No. H13701G

Chemical Identifiers

CAS 17334-08-6
Molecular Formula C5H8N2O
Molecular Weight (g/mol) 112.132
MDL Number MFCD00964673
InChI Key CDQDMLWGTVLQEE-UHFFFAOYSA-N
Synonym 1-methyl-1h-imidazol-2-yl methanol, 1-methylimidazol-2-yl methanol, 1h-imidazole-2-methanol, 1-methyl, 1-methyl-2-imidazolyl methanol, 2-hydroxymethyl-1-methylimidazole, imidazole-2-methanol, 1-methyl, 1-methyl-1h-imidazole-2-methanol, 1-methylimidazol-2-yl methan-1-ol, pubchem9047
PubChem CID 573612
IUPAC Name (1-methylimidazol-2-yl)methanol
SMILES CN1C=CN=C1CO

Specifications

Melting Point 117°C
Color White
Quantity 1 g
Formula Weight 112.13
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxymethyl-1-methylimidazole